Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

H6654

Sigma-Aldrich

Methyl 4-hydroxybenzoate

BioXtra, ≥99.0% (titration)

Synonym(s):

p-Hydroxybenzoic acid methyl ester, Methyl paraben, NIPAGIN

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CO2CH3
CAS Number:
Molecular Weight:
152.15
Beilstein:
509801
EC Number:
MDL number:
UNSPSC Code:
51452201
PubChem Substance ID:
NACRES:
NA.25

product line

BioXtra

Quality Level

Assay

≥99.0% (titration)

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.1%

mp

125-128 °C (lit.)

solubility

ethanol: 0.1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.001%
NH4+: ≤0.05%
Na: ≤0.02%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

COC(=O)c1ccc(O)cc1

InChI

1S/C8H8O3/c1-11-8(10)6-2-4-7(9)5-3-6/h2-5,9H,1H3

InChI key

LXCFILQKKLGQFO-UHFFFAOYSA-N

Gene Information

rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

334.4 °F

Flash Point(C)

168 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyung-Jin Min et al.
Aging cell, 7(2), 199-206 (2008-01-29)
Reduced insulin/insulin-like growth factor (IGF) signaling may be a natural way for the reduction of dietary nutrients to extend lifespan. While evidence challenging this hypothesis is accumulating with Caenorhabditis elegans, for Drosophila melanogaster it is still thought that insulin/IGF and
Ismail I Hewala et al.
Talanta, 88, 623-630 (2012-01-24)
Simple methods for HPTLC peak purity assessment and identification of the HPTLC peaks were presented. The spectrodensitograms - selected at different time intervals across the elution time of the HPTLC peak - were extracted and digital algorithms for manipulating the
Gabriela Oliveira et al.
International journal of pharmaceutics, 435(1), 38-49 (2012-05-29)
Simple topical formulations which include volatile components, such as gels or sprays, are appealing from a cosmetic perspective. However, complex formulation effects may result from the use of volatile excipients in topical formulations, particularly when applied at clinically relevant doses
Aaron Tustin et al.
Retina (Philadelphia, Pa.), 34(5), 935-942 (2013-10-10)
To determine whether the application of subconjunctival 2% lidocaine/0.1% methylparaben for anesthesia may reduce rates of endophthalmitis after intravitreal (IVT) injection. We performed in vitro experiments to determine the antibacterial properties of 2% lidocaine/0.1% methylparaben (lidocaine) against causative organisms of
Emilija Makaji et al.
International journal of toxicology, 30(4), 410-418 (2011-06-28)
There is an increasing concern that chemicals in the environment are contributing to the global rise in the prevalence of type 2 diabetes (T2D). However, there is limited evidence for direct effects of these chemicals on beta cell function. Therefore

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service