Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

H2910

Sigma-Aldrich

Hydrochlorothiazide

meets USP testing specifications

Synonym(s):

6-Chloro-3,4-dihydro-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-dioxide, 6-Chloro-7-sulfamyl-3,4-dihydro-1,2,4-benzothiadiazine 1,1-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8ClN3O4S2
CAS Number:
Molecular Weight:
297.74
EC Number:
MDL number:
UNSPSC Code:
12352125
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
meets USP testing specifications

Quality Level

form

solid

application(s)

pharmaceutical (small molecule)

SMILES string

NS(=O)(=O)c1cc2c(NCNS2(=O)=O)cc1Cl

InChI

1S/C7H8ClN3O4S2/c8-4-1-5-7(2-6(4)16(9,12)13)17(14,15)11-3-10-5/h1-2,10-11H,3H2,(H2,9,12,13)

InChI key

JZUFKLXOESDKRF-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Carbonic anhydrase inhibitor

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A Dornhorst et al.
Lancet (London, England), 1(8421), 123-126 (1985-01-19)
14 hypertensive men with type II diabetes sequentially received, in random order, hydrochlorothiazide 50 mg twice a day, propranolol 80 mg twice a day, and both drugs in combination. The 3-week treatment periods were separated by a 1-week washout period.
Irfan A Dhalla et al.
Annals of internal medicine, 158(6), 447-455 (2013-04-05)
Some evidence suggests that chlorthalidone may be superior to hydrochlorothiazide for the treatment of hypertension. To compare the effectiveness and safety of chlorthalidone and hydrochlorothiazide in older adults. Propensity score-matched observational cohort study with up to 5 years of follow-up.
Franz H Messerli et al.
The American journal of medicine, 124(10), 896-899 (2011-10-04)
Hydrochlorothiazide (HCTZ) has become by far the most commonly prescribed antihypertensive drug in the US. In 2008, 47.8 million prescriptions were written for HCTZ alone and 87.1 million prescriptions for HCTZ combinations. However, there is no evidence that HCTZ in
George C Roush et al.
Hypertension (Dallas, Tex. : 1979), 59(6), 1110-1117 (2012-04-25)
Hydrochlorothiazide (HCTZ) is widely used for hypertension, and prescriptions for HCTZ outnumber those for chlorthalidone (CTDN) by >20-fold in 2 recent surveys. Some have recently expressed a preference for CTDN. However, head-to-head trials testing the effect of the 2 drugs
Johannes A N Dorresteijn et al.
Journal of hypertension, 31(2), 393-403 (2012-12-14)
The objective of this study is to determine the effects of renin-angiotensin-aldosterone system inhibition, sympathoinhibition and diuretic therapy on endothelial function and blood pressure in obesity-related hypertension. A randomized, four-way, double-blind, crossover study in 31 adults with previously untreated obesity-related

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service