Skip to Content
MilliporeSigma
All Photos(1)

Documents

H0830000

Homatropine methylbromide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Homatropine methobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H24BrNO3
CAS Number:
Molecular Weight:
370.28
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

homatropine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C17H24NO3.BrH/c1-18(2)13-8-9-14(18)11-15(10-13)21-17(20)16(19)12-6-4-3-5-7-12;/h3-7,13-16,19H,8-11H2,1-2H3;1H/q+1;/p-1

InChI key

FUFVKLQESJNNAN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Homatropine methylbromide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Milena V Deolindo et al.
The European journal of neuroscience, 38(3), 2374-2381 (2013-05-15)
The aim of the present study was to investigate the role of the lateral hypothalamus (LH) and its local glutamatergic neurotransmission in the cardiovascular adjustments observed when rats are submitted to acute restraint stress. Bilateral microinjection of the nonspecific synaptic
Application of second-derivate UV spectrophotometry to the determination of homatropine methylbromide in single-component dosage form.
M Knochen et al.
Bollettino chimico farmaceutico, 126(7), 294-297 (1987-07-01)
Litao Yang et al.
Journal of gastroenterology and hepatology, 28(10), 1648-1653 (2013-06-05)
Hepatic angiomyolipoma (AML) is a rare, hepatic mesenchymal neoplasm. Its preoperative diagnosis is very difficult, and the treatment is still controversial. The aim is to summarize experience in diagnosis and management of hepatic AML from a cancer center. We retrospectively
J A Ramírez-Mayans et al.
Boletin medico del Hospital Infantil de Mexico, 46(12), 816-824 (1989-12-01)
In this article, the neurophysiology and neurobiochemistry of the digestive system are reviewed briefly. The pharmacology of the main gastrointestinal prokinetic drugs and their indications, side effects and dosages in pediatric patients are described.
Leonardo B M Resstel et al.
European journal of pharmacology, 580(3), 372-379 (2007-12-18)
We have previously reported that l-glutamate (L-glu) injected into the ventral portion of medial prefrontal cortex (vMPFC) of unanesthetized normotensive Wistar rats elicited cardiovascular responses. In the present study we investigated whether the spontaneously hypertensive rat (SHR) exhibit abnormal cardiovascular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service