Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F5422

Millipore

Folic acid assay medium

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171608
eCl@ss:
34058014
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

pH

6.8±0.2 (25 °C)

application(s)

food and beverages
pharmaceutical

microbiology

storage temp.

2-8°C

suitability

nonselective for Streptococcus spp.

Application

Recommended for the microbiological assay of folic acid using Streptococcus faecium as the test organism.

Components

Components (g/L)
Casein Acid Hydrolysate, Vitamin Free 12.00
Dextrose 40.00
Sodium Citrate 20.00
L-Cystine 0.20
DL-Tryptophan 0.20
Adenine Sulfate 0.02
Guanine Hydrochloride 0.02
Uracil 0.02
Thiamine Hydrochloride 0.002
Pyridoxine Hydrochloride 0.004
Riboflavin (Vitamin B2) 0.002
Nacin 0.002
p-Aminobenzoic Acid (PABA) 0.0002
Biotin 0.0000008
Calcium Pantothenate 0.0004
Dipotassium Phosphate 1.00
Monopotassium Phosphate 1.00
Magnesium Sulfate 0.40
Sodium Chloride 0.02
Ferrous Sulphate 0.02
Manganese Sulphate 0.02

Preparation Note

Suspend 7.5 g of Folic Acid Assay Medium in 100 ml of distilled water. Boil to dissolve the medium completely. Mix well to distribute the slight precipitate evenly. For assay, dispense 5 ml of medium per assay tube, and add 5 ml of distilled water to each tube. Sterilize by autoclaving at 15 lbs. pressure (121°C) for 10 minutes. Cool immediately.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service