Skip to Content
MilliporeSigma

F2270

Biofreeze Freezing Medium

DMSO-free, Serum-free, Animal Component-free Cryopreservation Medium, 1X, sterile-filtered, suitable for cell culture

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.71
UNSPSC Code:
12352207
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

sterility

sterile-filtered

technique(s)

cell culture | mammalian: suitable

shipped in

ambient

storage temp.

2-8°C



Storage Class

10 - Combustible liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library