Skip to Content
MilliporeSigma

F1628

Fluorescein isothiocyanate isomer I–Celite®

8.0 - 12.0% purity, powder

Synonym(s):

FITC–Celite®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Fluorescein isothiocyanate isomer I–Celite®, suitable for fluorescent labeling techniques

form

powder

Quality Segment

extent of labeling

8-12% FITC

color

yellow to orange

suitability

suitable for fluorescent labeling techniques

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

Oc1ccc2c(Oc3cc(O)ccc3C24OC(=O)c5cc(ccc45)N=C=S)c1

InChI

1S/C21H11NO5S/c23-12-2-5-16-18(8-12)26-19-9-13(24)3-6-17(19)21(16)15-4-1-11(22-10-28)7-14(15)20(25)27-21/h1-9,23-24H

InChI key

MHMNJMPURVTYEJ-UHFFFAOYSA-N

Application

Fluorescein isothiocyanate isomer I–Celite® has been conjugated with antibody for fluorescence imaging in cells and flow cytometry analysis.

Legal Information

Celite is a registered trademark of Imerys Minerals California, Inc.


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library