Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F0550000

Fructose

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

D-(−)-Fructose, D-Levulose, Fruit sugar

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1239004
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

fructose

manufacturer/tradename

EDQM

mp

119-122 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3,5-9,11-12H,1-2H2/t3-,5-,6-/m1/s1

InChI key

BJHIKXHVCXFQLS-UYFOZJQFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fructose EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jérémy Lothier et al.
Planta, 240(3), 629-643 (2014-07-16)
The first 6-fructan exohydrolase (6-FEH) cDNA from Lolium perenne was cloned and characterized. Following defoliation, Lp6 - FEHa transcript level unexpectedly decreased together with an increase in total FEH activity. Lolium perenne is a major forage grass species that accumulates
M Li et al.
The Journal of nutritional biochemistry, 26(3), 267-276 (2015-01-13)
Excessive fructose consumption is associated with insulin resistance (IR) and nonalcoholic fatty liver disease (NAFLD), and high fructose intake during pregnancy can lead to compromised fetal development in the rat. Evidence suggests that the amino acid taurine can ameliorate fructose-induced
Carlos A Roncal Jimenez et al.
Kidney international, 86(2), 294-302 (2013-12-18)
The epidemic of chronic kidney disease in Nicaragua (Mesoamerican nephropathy) has been linked with recurrent dehydration. Here we tested whether recurrent dehydration may cause renal injury by activation of the polyol pathway, resulting in the generation of endogenous fructose in
Thing-Fong Tzeng et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 69, 132-139 (2014-04-09)
We investigated the effects of zerumbone, a natural cyclic sesquiterpene of Zingiber zerumbet Smith, on high-fat diet (HFD)-induced hyperlipidemic hamsters. After being fed HFD for 2 weeks, Syrian golden hamsters were dosed orally with zerumbone (25, 50, and 100 mg/kg)
Yu Wang et al.
Journal of agricultural and food chemistry, 62(25), 5925-5931 (2014-05-27)
To evaluate the activity of botanicals used in Chinese Traditional Medicine as hypoglycemic agents for diabetes type II prevention and/or treatment, extracts prepared from 26 medicinal herbs were screened for their inhibitory activity on sodium-dependent glucose transporter 1 (SGLT1) by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service