Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ERMEF318K

Gas-Oil (high content Solvent Yellow 124)

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

application(s)

petroleum

format

matrix material

Legal Information

ERM is a registered trademark of European Commission

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 3 - STOT RE 2 Inhalation

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

>132.8 °F

Flash Point(C)

> 56 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service