Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.24
UNSPSC Code:
41116107
grade
certified reference material
agency
ERM®
manufacturer/tradename
JRC
application(s)
petroleum
format
matrix material
Analysis Note
A JRC Certificate of Analysis is included with the product, a pdf copy of the CoA and other documentation you can find here:
please visit the JRC webpage
please visit the JRC webpage
Legal Information
ERM is a registered trademark of European Commission
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Asp. Tox. 1 - Carc. 1B - Flam. Liq. 2 - Muta. 1B
Storage Class
3 - Flammable liquids
Flash Point (°F)
71.6 °F
Flash Point (°C)
22 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

