Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ERMEC591

Polypropylene (PP)

ERM®, certified reference material

Synonym(s):

Polypropylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(CH3)]n
CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

density

0.9 g/mL at 25 °C (lit.)

application(s)

chemicals and industrial polymers

format

neat

storage temp.

−20°C

SMILES string

CC=C

InChI

1S/C22H42O3/c1-2-3-4-5-11-14-17-20-21(25-20)18-15-12-9-7-6-8-10-13-16-19-22(23)24/h20-21H,2-19H2,1H3,(H,23,24)/t20-,21+/m1/s1

InChI key

NSYDMBURIUSUDH-RTWAWAEBSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
ERMEC591

Legal Information

ERM is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Thomas P J Linsinger et al.
Analytical chemistry, 81(10), 3792-3800 (2009-03-28)
The first reference materials certified for several polybrominated flame retardants in polymers were developed. Commercially available polyethylene and polypropylene were fortified with technical mixtures of Pentabrominated diphenylether (Penta-BDE), Octa-BDE, Deca-BDE, and Decabrominated biphenyl (BB) (where the capitalized forms refer to
G Imanidis et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 47(3), 283-287 (1999-06-26)
The possibility to control the release rate of salbutamol through the hydrophobic Celgard 2500 polypropylene membrane by varying the composition and the viscosity of hydrophilic drug vehicles was investigated. The use of the polypropylene membrane as a control membrane for
Simon J Gallagher et al.
Journal of drug targeting, 11(6), 373-379 (2003-12-12)
The aim of this study was to test the hypothesis that the most appropriate model for studying the diffusional release of an active from a topical formulation is one in which the membrane offers minimal resistance to release and involves
F Cassiola et al.
Artificial organs, 24(3), 168-173 (2000-04-12)
The introduction of microporous polypropylene hollow fibers in recent years has brought considerable advances to blood oxygenators. However, lifetime and assembly problems are still unresolved. In this work we tried to rate the oxygen permeation velocity by turning the fibers
[Procedure for membrane oxygenation of blood over hydrophilic polymer membranes].
H Körber et al.
Biomedizinische Technik. Biomedical engineering, 43 Suppl, 338-340 (1998-12-22)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service