Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D3281

Supelco

Diflunisal

analytical standard

Synonym(s):

2′,4′-Difluoro-4-hydroxy-[1,1′-biphenyl]-3-carboxylic acid, 5-(2,4-Difluorophenyl)salicylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8F2O3
CAS Number:
Molecular Weight:
250.20
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

FC1=CC(F)=CC=C1C2=CC(C(O)=O)=C(O)C=C2

InChI

1S/C13H8F2O3/c14-8-2-3-9(11(15)6-8)7-1-4-12(16)10(5-7)13(17)18/h1-6,16H,(H,17,18)

InChI key

HUPFGZXOMWLGNK-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John L Berk et al.
Amyloid : the international journal of experimental and clinical investigation : the official journal of the International Society of Amyloidosis, 19 Suppl 1, 37-38 (2012-05-04)
Familial amyloidotic polyneuropathy (FAP) is a protein folding disorder that induces neuropathy and cardiomyopathy, leading to death within 7-15 years after onset of clinical disease. In vitro, small ligands binding the thyroid hormone docking site stabilize tetrameric transthyretin, inhibiting amyloid
Teresa Mairal et al.
PloS one, 4(1), e4124-e4124 (2009-01-07)
The thyroid hormone and retinol transporter protein known as transthyretin (TTR) is in the origin of one of the 20 or so known amyloid diseases. TTR self assembles as a homotetramer leaving a central hydrophobic channel with two symmetrical binding
Maria Esther Zugasti et al.
Drug development and industrial pharmacy, 35(10), 1264-1270 (2009-06-27)
The influence of beta-cyclodextrin (beta-CD) polymers on drug release from hydroxypropyl methylcellulose (HPMC) matrices has not been reported in the literature. The influence of monomeric beta-CD and both soluble and insoluble beta-CD polymers on drug release from tablets containing either
Stella Fountoulaki et al.
Journal of inorganic biochemistry, 105(12), 1645-1655 (2011-11-11)
Copper(II) complexes with the non-steroidal anti-inflammatory drug diflunisal in the presence of N,N-dimethylformamide or nitrogen donor heterocyclic ligands (pyridine, 1,10-phenanthroline, 2,2'-bipyridine or 2,2'-bipyridylamine) have been synthesized and characterized. The deprotonated diflunisal ligands are coordinated to Cu(II) ion through carboxylato oxygen
J A Murillo Pulgarín et al.
Analytica chimica acta, 625(1), 47-54 (2008-08-30)
The simultaneous determination of 6-methoxy-2-naphthylacetic acid (6MNA) and diflunisal in serum samples using the combination of matrix isopotential synchronous fluorescence (MISF) and first derivative technique is proposed. 6MNA and diflunisal exhibit overlapped spectra and serum produces background fluorescence that precludes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service