Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLNSOIL2

Supelco

Clean Clay Loam

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

CofA

current certificate can be downloaded

manufacturer/tradename

RTC CLNSOIL-2-100

matrix

soil

application(s)

environmental

format

matrix material

storage temp.

2-30°C

General description

Clean clay loam may be used as a reference material in the recovery of nitro-organic explosives in soil samples using gas chromatography coupled with electron capture detector (GC-ECD).

Application

Product is intended for use as a clean control and as a base for fortification when no suitable CRM is available for use in method validation, method development or matrix recovery applications. COA covers most paramaters of interest.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of a solid phase extraction sample cleanup procedure for the recovery of trace levels of nitro-organic explosives in soil.
Thomas L J, et al.
Forensic Science International (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service