Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

C9606

Sigma-Aldrich

Castor oil

meets testing specifications per USP

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.21

biological source

Ricinus communis

Quality Level

Agency

USP/NF
meets USP testing specifications

form

viscous liquid

quality

meets testing specifications per USP

refractive index

n20/D 1.478 (lit.)

bp

313 °C (lit.)

density

0.961 g/cm3 at 25 °C

lipid type

oils

shipped in

ambient

storage temp.

room temp

SMILES string

O(C(COC(=O)CCCCCCC\C=C/CC(O)CCCCCC)COC(=O)CCCCCCC\C=C/CC(O)CCCCCC)C(=O)CCCCCCC\C=C/CC(O)CCCCCC

InChI key

ZEMPKEQAKRGZGQ-AAKVHIHISA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Selection of lubricant type and concentration for orodispersible tablets: This study evaluates the efficacy of various lubricants, including castor oil, for use in the pharmaceutical industry, specifically in the manufacturing of orodispersible tablets (Yi Zheng A et al., 2024) (doi: 10.1016/j.ijpharm.2024.124190).

  • Polymers from Plant Oils Linked by Siloxane Bonds for Programmed Depolymerization: Focuses on the synthesis of innovative polymers from castor oil, linked via siloxane bonds, aimed at enhancing the sustainability of material production through easier depolymerization (Cheng C et al., 2024) (doi: 10.1021/jacs.4c01982).

  • Mechanical properties and curing kinetics of bio-based benzoxazine-epoxy copolymer for dental fiber post: Explores the use of castor oil in the creation of bio-based benzoxazine-epoxy copolymers, focusing on their mechanical properties and curing kinetics, with applications in dental materials (Mora P et al., 2023) (doi: 10.1186/s40643-023-00684-x).

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bruno Rodrigues Sampieri et al.
Veterinary parasitology, 191(3-4), 315-322 (2012-10-09)
Rhipicephalus sanguineus are bloodsucking ectoparasites, whose main host is the domestic dog, thus being present in urban areas and closely located to people. Eventually, this tick species parasitize humans and can become a potential vector of infectious diseases. Methods to
Hypersensitivity reaction to intravenous but not oral tacrolimus.
Sarah Nicolai et al.
Transplantation, 94(9), e61-e63 (2012-11-07)
S Rajan et al.
Asian Pacific journal of tropical medicine, 5(8), 630-633 (2012-07-31)
To examine the antidiarrhoeal activity of alcoholic and aqueous seed kernel extract of Mangifera indica (M. indica) on castor oil-induced diarrhoeal activity in Swiss albino mice. Mango seed kernels were processed and extracted using alcohol and water. Antidiarrhoeal activity of
Alejandro Ruiz Olivares et al.
Journal of environmental management, 114, 316-323 (2012-11-23)
Bioenergy production combined with phytoremediation has been suggested to help in solving two critical world problems: the gradual reduction of fossil fuels and soil contamination. The aim of this research was to investigate the potential for the use of Ricinus
E C Alonso et al.
Journal of economic entomology, 106(2), 742-746 (2013-06-22)
Leaf-cutting ants are the main herbivores in the New World tropics. Although the toxicity of seed oils against these ants has been poorly investigated, previous results revealed that seed oils exert considerable toxic activity against these insects. This paper analyzes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service