Skip to Content
MilliporeSigma

C3607

Cyanocobalamin

meets USP testing specifications

Synonym(s):

Vitamin B12, α-(5,6-Dimethylbenzimidazolyl)cyanocobamide, CN-Cbl, Cyanocob(III)alamin, Cyanocobalamin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C63H88CoN14O14P
CAS Number:
Molecular Weight:
1355.37
MDL number:
UNSPSC Code:
12352205
eCl@ss:
34058004
PubChem Substance ID:
EC Number:
200-680-0
NACRES:
NA.21
Beilstein/REAXYS Number:
4122889
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

USP/NF, meets USP testing specifications

assay

96.0-102.0% dry basis

form

powder

color

dark red

application(s)

pharmaceutical (small molecule)

storage temp.

2-8°C

SMILES string

C/C(C1=N[C@]([C@]([C@H](CC(N)=O)[C@@]/2(C)CCC(NC[C@@H](OP([O-])(O[C@H]3[C@@H](O)[C@@H](N4C=[N]5C6=C4C=C(C)C(C)=C6)O[C@@H]3CO)=O)C)=O)([H])N([Co+]5C#N)C2=C(C)/C([C@@H](CCC(N)=O)C/7(C)C)=NC7=C/8)(C)[C@@](C)(CC(N)=O)[C@@H]1CCC(N)=O)=C9N=C8[C@@H](CCC(N)=O)[C@

InChI

1S/C62H90N13O14P.CN.Co/c1-29-20-39-40(21-30(29)2)75(28-70-39)57-52(84)53(41(27-76)87-57)89-90(85,86)88-31(3)26-69-49(83)18-19-59(8)37(22-46(66)80)56-62(11)61(10,25-48(68)82)36(14-17-45(65)79)51(74-62)33(5)55-60(9,24-47(67)81)34(12-15-43(63)77)38(71-55)23-42-58(6,7)35(13-16-44(64)78)50(72-42)32(4)54(59)73-56;1-2;/h20-21,23,28,31,34-37,41,52-53,56-57,76,84H,12-19,22,24-27H2,1-11H3,(H15,63,64,65,66,67,68,69,71,72,73,74,77,78,79,80,81,82,83,85,86);;/q;;+2/p-2/t31-,34-,35-,36-,37+,41-,52-,53-,56-,57?,59-,60+,61+,62+;;/m1../s1

InChI key

RMRCNWBMXRMIRW-QJRSUKKJSA-L

General description

Cyanocobalamin (B12) is a water soluble vitamin, which is mainly found in animal products, like dairy products, meat and eggs. It belongs to the B12 class of vitamins, commonly called as cobalamins. Cyanocobalamin consists of four pyrrole rings with a central cobalt atom coordinated by four equatorial nitrogen ligands.

Application

Required for growth, genetic stability and survival of cells in vitro. Functions to support one-carbon metabolism. Present in many classical and serum-free formulations.
Cyanocobalamin has been used to study its effects on the prevention of colon cancer.

Biochem/physiol Actions

Cyanocobalamin is used in the preparation of multivitamin supplements and fortified food. It acts as a coenzyme to convert L-methylmalonyl CoA to succinyl CoA, which is catalyzed by isomerase. Cyanocobalamin is used to treat vitamin B12 deficiency. Cyanocobalamin meets USP testing specifications.


Still not finding the right product?

Explore all of our products under Cyanocobalamin


Storage Class

11 - Combustible Solids

ppe

Eyeshields, Gloves, type N95 (US)

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library