Skip to Content
MilliporeSigma

C0501

Casein Acid Hydrolysate for Biotechnology and Microbiology

for Biotechnology and Microbiology, ≥7.8% Total N, 5.2-7.7% Amino

Synonym(s):

Casein hydrolysate, Hy-Case® Amino

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106212
NACRES:
NA.74
EC Number:
265-363-1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Hy-Case® Amino Casein Acid Hydrolysate, for Biotechnology and Microbiology

biological source

bovine milk

Quality Segment

form

powder

contains

25-40% sodium chloride

packaging

pkg of (1 kg in poly bottle
100, 500 g in poly bottle)

nitrogen analysis

≥7.8% total

loss

≤4% loss on drying

pH

4.7-6.7

application(s)

biotechnology/fermenter
microbiology
microbiology

General description

Hy-Case® Amino is an acid digest of casein processed to a high degree of hydrolysis. It is a refined hydrolysate with high solubility and is used as a microbiological nutrient in laboratory media and fermentations. Casein hydrolysate is produced by the digestion of casein with hydrochloric acid. The digestion conditions are such that a proportion of vitamins and growth promoting substances are retrained. Tryptophane is destroyed by the acid treatment. The inorganic salts level is high due to neutralization of the acid.

Application

Hy-Case® Amino or Casein hydrolysate is especially suited for the large-scale cultivation of Streptococci, for toxins and streptokinase production. In industrial fermentations it favors a high yield of biomass, particularly for certain yeasts and fastidious bacilli. It is also used as a nutrient in vaccine and toxin production where a high free amino acid content is required.

Preparation Note

Recommended concentration: 150 g/L

Analysis Note

Primarily free amino acids with a small amount of peptides.

Legal Information

Hy-Case is a registered trademark of Kerry Group


Still not finding the right product?


Storage Class

11 - Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library