Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C0350000

Calcium oxalate monohydrate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Oxalic acid calcium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(COO)2Ca · H2O
CAS Number:
Molecular Weight:
146.11
Beilstein:
3757668
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Ca++].[H]O[H].[O-]C(=O)C([O-])=O

InChI

1S/C2H2O4.Ca.H2O/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);;1H2/q;+2;/p-2

InChI key

LQHWSGSWNOHVHO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Calcium oxalate monohydrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haoran Liu et al.
Theranostics, 10(16), 7319-7334 (2020-07-10)
Intrarenal calcium oxalate (CaOx) crystals induce renal tubular epithelial cells (TECs) injury and inflammation, which involve Toll-like receptor 4 (TLR4)/interferon regulatory factor 1 (IRF1) signaling. Additionally, infiltrating macrophages (Mϕs) might influence intrarenal CaOx crystals and CaOx-induced renal injury. Although the
Zhu Wang et al.
Urolithiasis, 47(3), 225-234 (2018-06-28)
To study the expression profiles of lncRNA and mRNA in the calcium oxalate monohydrate-attached HK-2 cells, and investigate the association between critical lncRNA expression level and renal injury. The HK-2 cells were treated with crystal suspension of calcium oxalate. The
Jonathan D Harper et al.
The Journal of urology, 190(3), 1090-1095 (2013-04-16)
Focused ultrasound has the potential to expel small stones or residual stone fragments from the kidney, or move obstructing stones to a nonobstructing location. We evaluated the efficacy and safety of ultrasonic propulsion in a live porcine model. Calcium oxalate
Elaine M Worcester et al.
American journal of physiology. Renal physiology, 305(11), F1574-F1584 (2013-10-04)
The sequence of events by which primary hyperoxaluria type 1 (PH1) causes renal failure is unclear. We hypothesize that proximal tubule (PT) is vulnerable because oxalate secretion raises calcium oxalate (CaOx) supersaturation (SS) there, leading to crystal formation and cellular
B Hess
Urological research, 20(1), 83-86 (1992-01-01)
The processes of calcium oxalate monohydrate (COM) crystal nucleation, growth and aggregation (agglomeration) generally have been studied using a wide variety of assay systems/conditions. This paper reviews the apparently conflicting data on the effects of Tamm-Horsfall glycoprotein (THP) on COM

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service