Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BP528

Lormetazepam

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

N-Methyllorazepam

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12Cl2N2O2
CAS Number:
Molecular Weight:
335.18
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

lormetazepam

manufacturer/tradename

BP

drug control

psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

format

neat

storage temp.

2-8°C

SMILES string

Clc1c(cccc1)C2=NC(C(=O)N(c3c2cc(cc3)Cl)C)O

InChI

1S/C16H12Cl2N2O2/c1-20-13-7-6-9(17)8-11(13)14(19-15(21)16(20)22)10-4-2-3-5-12(10)18/h2-8,15,21H,1H3

InChI key

FJIKWRGCXUCUIG-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lormetazepam BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • SC VI A. Pharmacopoeial Calculations
  • Lormetazepam Tablets
  • Lormetazepam
  • Packaging

    Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    A López-Rabuñal et al.
    Journal of analytical toxicology, 44(6), 580-588 (2020-02-18)
    An LC-MS-MS method for the determination of 14 benzodiazepines (BZDs) (alprazolam, α-hydroxyalprazolam, clonazepam, bromazepam, diazepam, nordiazepam, lorazepam, lormetazepam, oxazepam, flunitrazepam, 7-aminoflunitrazepam, triazolam, midazolam and zolpidem) and 15 antidepressants (ADs) (amitriptyline, nortriptyline, imipramine, desipramine, clomipramine, norclomipramine, fluoxetine, norfluoxetine, sertraline, norsertraline, paroxetine
    Satomi Kagota et al.
    European journal of pharmacology, 892, 173744-173744 (2020-11-22)
    Benzodiazepines (BDZs) and non-BDZ sedative-hypnotics are effective for the management of chronic insomnia; however, they are associated with adverse effects such as headache, dizziness, and palpitations. Furthermore, long-term use of these medications is associated with decreased blood pressure (BP) or
    Alicja Majda et al.
    Talanta, 209, 120533-120533 (2020-01-02)
    For the first time the method DI-SPME/LC-TOFMS was used and developed in order to determine the large antidepressant drugs in real forensic cases. The aim of the study was to optimize the new DI-SPME/LC-TOFMS method for the quantification of the

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service