Skip to Content

BP488

Erythromycin stearate

British Pharmacopoeia (BP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C37H67NO13 · C18H36O2
CAS Number:
Molecular Weight:
1018.40
UNSPSC Code:
41116107
NACRES:
NA.24
EC Number:
211-396-1
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

erythromycin

form

solid

quality

British Pharmacopoeia (BP) Reference Standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCC(O)=O.CC[C@@H]1OC(=O)[C@@H](C)[C@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@@H](C)[C@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@@](C)(O)C[C@H](C)C(=O)C(C)[C@@H](O)[C@]1(C)O

InChI

1S/C37H67NO13.C18H36O2/c1-14-25-37(10,45)30(41)20(4)27(39)18(2)16-35(8,44)32(51-34-28(40)24(38(11)12)15-19(3)47-34)21(5)29(22(6)33(43)49-25)50-26-17-36(9,46-13)31(42)23(7)48-26;1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h18-26,28-32,34,40-42,44-45H,14-17H2,1-13H3;2-17H2,1H3,(H,19,20)/t18-,19+,20?,21+,22-,23-,24-,25-,26-,28+,29+,30+,31-,32-,34-,35-,36+,37?;/m0./s1

InChI key

YAVZHCFFUATPRK-JGJPHTNUSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Erythromycin stearate BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Erythromycin Stearate Tablets
  • Packaging

    Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.


    Still not finding the right product?

    Explore all of our products under Erythromycin stearate


    pictograms

    Health hazard

    signalword

    Danger

    hcodes

    Hazard Classifications

    Resp. Sens. 1 - Skin Sens. 1

    Storage Class

    11 - Combustible Solids

    wgk

    WGK 2

    flash_point_f

    Not applicable

    flash_point_c

    Not applicable



    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Don't see the Right Version?

    If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library