Skip to Content
MilliporeSigma

BP400

Rifampicine N-oxide

British Pharmacopoeia (BP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C43H58N4O13
CAS Number:
Molecular Weight:
838.94
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
eCl@ss:
25140904
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

rifampicin

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

[N+](=C\c2c3c(c4c(c5c(c(c4O)C)O[C@](O\C=C\[C@@H]([C@H]([C@H]([C@@H]([C@@H]([C@@H]([C@H]([C@H](\C=C\C=C(/C(=O)N3)\C)C)O)C)O)C)OC(=O)C)C)OC)(C5=O)C)c2O)O)(/N1CCN(CC1)C)\[O-]

InChI

1S/C43H58N4O13/c1-21-12-11-13-22(2)42(55)44-33-28(20-47(56)46-17-15-45(9)16-18-46)37(52)30-31(38(33)53)36(51)26(6)40-32(30)41(54)43(8,60-40)58-19-14-29(57-10)23(3)39(59-27(7)48)25(5)35(50)24(4)34(21)49/h11-14,19-21,23-25,29,34-35,39,49-53H,15-18H2,1-10H3,(H,44,55)/b12-11+,19-14+,22-13-,47-20-/t21-,23+,24+,25+,29-,34-,35+,39+,43-/m0/s1

InChI key

PADXLTATLMUKBQ-RIPAWWNWSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Rifampicine N-oxide BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Packaging

Unit quantity: 25 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

Other Notes

Sales restrictions may apply.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library