Skip to Content
MilliporeSigma

Skip To

BP024

2-Azahypoxanthine

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

3,7-Dihydro-4H-imidazo[4,5-d]-1,2,3-triazin-4-one, 2-Azahypoxanthine, NSC 22419

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C4H3N5O
CAS Number:
Molecular Weight:
137.10
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

dacarbazine

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

N1NC(=O)C2=NC=NC2=N1

InChI

1S/C4H3N5O/c10-4-2-3(6-1-5-2)7-9-8-4/h1,9H,(H,8,10)

InChI key

WXKVKFJUCMCVFI-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support

Application

2-Azahypoxanthine BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Dacarbazine Injection
  • Packaging

    Unit quantity: 50 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    Compare Similar Items

    View Full Comparison

    Show Differences

    1 of 1

    This Item
    BP010BP001BP046
    manufacturer/tradename

    BP

    manufacturer/tradename

    BP

    manufacturer/tradename

    BP

    manufacturer/tradename

    BP

    grade

    pharmaceutical primary standard

    grade

    pharmaceutical primary standard

    grade

    pharmaceutical primary standard

    grade

    pharmaceutical primary standard

    format

    neat

    format

    neat

    format

    neat

    format

    neat

    application(s)

    pharmaceutical
    pharmaceutical small molecule

    application(s)

    pharmaceutical
    pharmaceutical small molecule

    application(s)

    pharmaceutical
    pharmaceutical small molecule

    application(s)

    pharmaceutical
    pharmaceutical small molecule

    shelf life

    limited shelf life, expiry date on the label

    shelf life

    limited shelf life, expiry date on the label

    shelf life

    limited shelf life, expiry date on the label

    shelf life

    limited shelf life, expiry date on the label

    form

    solid

    form

    crystalline

    form

    solid

    form

    solid


    Storage Class

    11 - Combustible Solids

    wgk

    WGK 3

    flash_point_f

    Not applicable

    flash_point_c

    Not applicable



    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library



    Questions

    Reviews

    No rating value

    Active Filters