Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BCR377

Maize flour (deoxynivalenol, blank)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

application(s)

agriculture
cosmetics
food and beverages
pharmaceutical (small molecule)

format

matrix material

shipped in

wet ice

storage temp.

−20°C

Analysis Note

For more information please see:
BCR377

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Biotoxins / Mycotoxins: Deoxynivalenol

Matrix Group: Cereals, Bread, Rice and Maize

Check out our entire range of trichothecene type mycotoxin reference materials

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service