Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B0990000

Betahistine mesilate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Betahistine mesylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N2 · 2CH4O3S
CAS Number:
Molecular Weight:
328.41
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

betahistine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C8H12N2.2CH4O3S/c1-9-7-5-8-4-2-3-6-10-8;2*1-5(2,3)4/h2-4,6,9H,5,7H2,1H3;2*1H3,(H,2,3,4)

InChI key

ZBJJDYGJCNTNTH-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Betahistine mesilate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Gürkov et al.
European archives of oto-rhino-laryngology : official journal of the European Federation of Oto-Rhino-Laryngological Societies (EUFOS) : affiliated with the German Society for Oto-Rhino-Laryngology - Head and Neck Surgery, 270(4), 1231-1235 (2012-07-05)
This study aimed to assess whether standard-dose Betahistine (48 mg daily) exerts an effect upon the degree of endolymphatic hydrops in patients with Menière's disease using a retrospective case series in the setting of a tertiary neurotology referral centre. In
Isilay Oz et al.
ORL; journal for oto-rhino-laryngology and its related specialties, 75(4), 211-220 (2013-08-01)
The aim of this study was to evaluate the effect of combined hearing and tinnitus masking devices that are appropriately programmed for acoustic stimulations using wide-band noise over the specific frequency range of tinnitus. A total of 21 patients were
Adriana Pontin Garcia et al.
Brazilian journal of otorhinolaryngology, 79(3), 366-374 (2013-06-08)
Virtual reality technology can provide a wide range of sensory stimuli to generate conflicts of varying degrees of complexity in a safe environment. To verify the effect of a virtual realitybased balance rehabilitation program for patients with Menière's disease. This
Looks vestibular: irrational prescribing of antivertiginous drugs for older dizzy patients in general practice.
Otto R Maarsingh et al.
The British journal of general practice : the journal of the Royal College of General Practitioners, 62(603), 518-520 (2012-12-26)
[Efficacy of vestibo in the treatment of vertebrobasilar insufficiency].
E I Chukanova et al.
Zhurnal nevrologii i psikhiatrii imeni S.S. Korsakova, 112(4), 53-56 (2012-07-20)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service