Skip to Content
MilliporeSigma

A7534

Bovine Serum Albumin solution

35% in saline, contains azide, aseptically filled

Synonym(s):

Albumin solution from bovine serum, BSA solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.27
MDL number:
eCl@ss:
32160409
Biological source:
bovine
Assay:
33.0-37.4 protein basis (biuret)
Form:
solution
Technique(s):
ELISA: suitable, electrophoresis: suitable, western blot: suitable
Concentration:
33.0-37.4 mg/mL Biuret, 35% in saline
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

biological source

bovine

sterility

aseptically filled

assay

33.0-37.4 protein basis (biuret)

form

solution

purified by

heat shock fractionation

contains

sodium azide as preservative

does not contain

stabilizer

origin

USA origin

concentration

33.0-37.4 mg/mL Biuret, 35% in saline

technique(s)

ELISA: suitable, electrophoresis: suitable, western blot: suitable

UniProt accession no.

foreign activity

BT virus, none detected, VSV virus, none detected

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)



Still not finding the right product?

Explore all of our products under Bovine Serum Albumin solution


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library