Skip to Content
MilliporeSigma

A7284

Bovine Serum Albumin solution

30%±2% in 0.85% sodium chloride, aseptically filled

Synonym(s):

Albumin solution from bovine serum, BSA solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.25
MDL number:
eCl@ss:
32160409
Biological source:
bovine
Assay:
28-32% protein basis (biuret)
Form:
solution
Technique(s):
ELISA: suitable
Concentration:
28-32% protein (biuret), 30%±2% in 0.85% sodium chloride
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine

Quality Segment

sterility

aseptically filled

assay

28-32% protein basis (biuret)

form

solution

purified by

heat shock fractionation

contains

0.1% sodium azide as preservative

does not contain

stabilizer

origin

USA origin

concentration

28-32% protein (biuret), 30%±2% in 0.85% sodium chloride

technique(s)

ELISA: suitable

UniProt accession no.

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

General description

Bovine serum albumin (BSA) is bound in a single chain cross-linked with 17 cysteine residues. It comprises three homologous, structurally different domains and two sub-domains each. This globular, α-helical, non-glycosylated protein is produced in the liver.

Application

Bovine Serum Albumin solution has been used:
  • to study the effects of plasma protein on sulfated glycosaminoglycans (sGAGs)
  • to histologically evaluate the implanted allogeneic mesenchymal stem cells (MSCs) alginate constructs
  • as a component of phosphate-buffered saline (PBS) to treat red blood cells (RBC) for sample preparation

Biochem/physiol Actions

Bovine Serum Albumin (BSA) is the most widely used blocking agent in enzyme-linked immunosorbent assay (ELISA). It is also the main constituent of cell culture media. BSA promotes the differentiation of human embryonic stem cells (hESC) differentiation and assists the circulation of drugs, hormones, and fatty acids.


Still not finding the right product?

Explore all of our products under Bovine Serum Albumin solution


Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library