Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A0718

Sigma-Aldrich

Aluminum chloride hexahydrate

meets EP, BP, USP testing specifications

Synonym(s):

AlCl3.6H2O, Aluminium trichloride hexahydrate, Aluminium(III) chloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AlCl3 · 6H2O
CAS Number:
Molecular Weight:
241.43
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.21

Agency

meets BP testing specifications
meets EP testing specifications
meets USP testing specifications

Quality Level

vapor pressure

1 mmHg ( 100 °C)

quality

meets EP, BP, USP testing specifications

reaction suitability

reagent type: catalyst
core: aluminum

solubility

water: soluble 50 mg/mL, clear, colorless

application(s)

pharmaceutical (small molecule)

SMILES string

Cl[Al](Cl)Cl.O.O.O.O.O.O

InChI

1S/Al.3ClH.6H2O/h;3*1H;6*1H2/q+3;;;;;;;;;/p-3

InChI key

JGDITNMASUZKPW-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application


  • Enhanced Chromium (VI) Removal by Micron-Scale Zero-Valent Iron Pretreated with Aluminum Chloride under Aerobic Conditions.: This research focused on the use of aluminum chloride hexahydrate in the pretreatment of micron-scale zero-valent iron for the removal of chromium (VI) under aerobic conditions. The findings indicate that aluminum chloride hexahydrate significantly improves the removal efficiency, providing a potential solution for environmental remediation efforts (Long et al., 2024).

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT RE 1

Target Organs

Lungs

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aisha Nasir Hashmi et al.
The International journal of neuroscience, 125(4), 277-287 (2014-05-16)
Aluminium metal (Al) has been implicated in the etiology of many neurodegenerative diseases, most commonly the Alzheimer's disease (AD). Al causes biochemical defects by affecting the neurotransmitters level, causes conformational changes in amyloid β protein and increases amyloid accumulation in
Ji-Won Jung et al.
ACS nano, 9(7), 6717-6727 (2015-06-02)
Amorphous silicon (a-Si) has been intensively explored as one of the most attractive candidates for high-capacity and long-cycle-life anode in Li-ion batteries (LIBs) primarily because of its reduced volume expansion characteristic (∼280%) compared to crystalline Si anodes (∼400%) after full
Dragana M Vucić et al.
Acta poloniae pharmaceutica, 70(6), 1021-1026 (2014-01-05)
Antibacterial and antioxidant activity, total phenolic and flavonoid concentrations of aqueous, ethanol and ethyl acetate extracts from the leaves and flowers of Erica herbacea L. were studied. In vitro antibacterial activity of the extracts was determined by macrodilution method. Minimum
Syed Husain Mustafa Rizvi et al.
PloS one, 9(5), e98409-e98409 (2014-06-01)
Aluminium (Al) is the third most abundant element in the earth's crust and its compounds are used in the form of house hold utensils, medicines and in antiperspirant etc. Increasing number of evidences suggest the involvement of Al+3 ions in
Qian Yee Ang et al.
Analytical and bioanalytical chemistry, 407(22), 6747-6758 (2015-07-15)
Molecular imprinting is an emerging technique to create imprinted polymers that can be applied in affinity-based separation, in particular, biomimetic sensors. In this study, the matrix of siloxane bonds prepared from the polycondensation of hydrolyzed tetraethoxysilane (TEOS) was employed as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service