Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

99310

Supelco

Calcium ionophore I - cocktail A

Selectophore

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
NACRES:
NB.61

product line

Selectophore

Quality Level

form

liquid

composition

Calcium ionophore I, 10.0 wt. % (21192)
2-Nitrophenyl octyl ether, 89.0 wt. % (73732)
Sodium tetraphenylborate, 1.0 wt. % (72018)

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples

Packaging

Clear borosilicate glass, high hydrolytic resistance (Type I)

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F. Lanter et al.
Analytica Chimica Acta, 135, 51-51 (1982)
Qi Wu et al.
Plant physiology, 183(3), 1331-1344 (2020-05-06)
Hydrogen gas (H2) has a possible signaling role in many developmental and adaptive plant responses, including mitigating the harmful effects of cadmium (Cd) uptake from soil. We used electrophysiological and molecular approaches to understand how H2 ameliorates Cd toxicity in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service