Skip to Content
MilliporeSigma
All Photos(1)

Documents

95451

Supelco

TOC System Suitability Test Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319

Agency

in accordance with USP (application)

shelf life

limited shelf life, expiry date on the label

contains

phosphoric acid as stabilizer

technique(s)

TOC: suitable

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

Application

not suitable for instruments using gold lined sample cells, e.g. Anatel A-643, Sievers Model 400 (US) or Model 2244 OL (Europe)

Components

kit contains TOC system suitability solution, TOC standard solution, TOC water

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service