Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

95212

Sigma-Aldrich

L-Ascorbic acid

tested according to Ph. Eur.

Synonym(s):

Acidum ascorbicum, L-Threoascorbic acid, Antiscorbutic factor, Vitamin C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
Beilstein:
84272
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic

Quality Level

Agency

USP/NF
tested according to Ph. Eur.

Assay

99.0-100.5%

form

solid

color

white to off-white

pH

1.0-2.5 (25 °C, 176 g/L in water)

mp

190-194 °C (dec.)

solubility

water: soluble 176 g/L at 20 °C

SMILES string

OC([C@]([C@@H](O)CO)([H])O1)=C(O)C1=O

InChI

1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2-,5+/m0/s1

InChI key

CIWBSHSKHKDKBQ-JLAZNSOCSA-N

Gene Information

human ... SLC23A2(9962)

Looking for similar products? Visit Product Comparison Guide

General description

L-Ascorbic acid is the most biologically active form of ascorbic acid. It is hydrophilic and an unstable compound. L-Ascorbic acid contains a six-carbon lactone produced by plants and some animal species but not by humans and other primates. Ascorbic acid, also referred to as Vitamin C, is a water-soluble vitamin. Ascorbic acid carries a neutral charge, which gets converted to ascorbate by protonation. Vitamin C is a part of citrus fruits, such as broccoli, strawberries, turnip.

Application


  • Targeted screening of multiple anti-inflammatory components from Chrysanthemi indici Flos by ligand fishing with affinity UF-LC/MS: This study investigates the application of L-Ascorbic acid in enhancing the screening of anti-inflammatory compounds, utilizing its properties to stabilize reactive samples in cell cultures (Huang et al., 2024).

  • Phenolic Content, Antioxidant Potential, and Antimicrobial Activity of Uvaria chamae (Annonaceae), a Food Plant from Burkina Faso: Demonstrates the use of L-Ascorbic acid in the evaluation of antioxidant properties of plant extracts, which is crucial in the pharmaceutical research of natural products (Kabore et al., 2024).

Biochem/physiol Actions

L-Ascorbic acid functions according to its oxido-reduction property. It is a co-factor for hydroxylation. The biochemical actions of L-ascorbic acid is dependent on the activity of monoxygenase. It is involved in carnitine, collagen synthesis and production of neurotransmitters. Vitamin C exhibits anti-oxidant properties. Vitamin C plays a role in providing protection against photoaging and anti-aging effect. Deficiency of vitamin C leads to scurvy, bleeding gums, poor wound healing, anemia and muscle degeneration.
Ascorbic Acid, also known as Vitamin C, is a six-carbon lactone produced by plants and some animal species but not by humans and other primates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Topical vitamin C and the skin: mechanisms of action and clinical applications
Al-Niaimi F and Chiang N Y Z
The Journal of Clinical and Aesthetic Dermatology, 10(7), 14-14 (2017)
Vitamin C in health and disease: its role in the metabolism of cells and redox state in the brain
Figueroa M R and Rivas A S
Frontiers in Physiology, 6(4), 397-397 (2015)
Vitamin C in disease prevention and cure: an overview
Chambial S, et al.
Indian Journal of Clinical Biochemistry : IJCB, 28(4), 314-328 (2013)
Takashi Yoshida et al.
Nature chemical biology, 2(11), 596-607 (2006-09-26)
Transient receptor potential (TRP) proteins form plasma-membrane cation channels that act as sensors for diverse cellular stimuli. Here, we report a novel activation mechanism mediated by cysteine S-nitrosylation in TRP channels. Recombinant TRPC1, TRPC4, TRPC5, TRPV1, TRPV3 and TRPV4 of
Christopher Grunseich et al.
Neurobiology of disease, 70, 12-20 (2014-06-14)
Spinal and bulbar muscular atrophy (SBMA, Kennedy's disease) is a motor neuron disease caused by polyglutamine repeat expansion in the androgen receptor. Although degeneration occurs in the spinal cord and muscle, the exact mechanism is not clear. Induced pluripotent stem

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service