Skip to Content
MilliporeSigma
All Photos(1)

Documents

939064

Sigma-Aldrich

Lithium hydroxide ChemBeads

new

Synonym(s):

LiOH ChemBeads

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
LiHO
CAS Number:
Molecular Weight:
23.95
MDL number:
UNSPSC Code:
12352100

description

Reagent Type: Inorganic salt

Quality Level

form

solid

composition

loading of base, 14-16 wt. %

reaction suitability

core: lithium

SMILES string

[Li+].[OH-]

InChI

1S/Li.H2O/h;1H2/q+1;/p-1

InChI key

WMFOQBRAJBCJND-UHFFFAOYSA-M

General description

Lithium hydroxide (LiOH) on glass beads. Lithium chloride solution in water on electrolysis forms LiOH. In respiratory apparatus and submarines, it is utilized to uptake carbon dioxide. A study on the redox mechanism of titanium dioxide (TiO2) using cyclic voltammetry, X-ray diffraction, X-ray photoelectron spectroscopy (XPS) and Fourier transform infrared spectroscopy (FTIR) in aqueous LiOH electrolyte has been reported.

Application

Lithium hydroxide has been used in the following processes: Synthesis of lithium-doped zinc oxide (ZnO) thin films. Preparation of lithium glyceroxide/hydroxide catalysts by reacting with glycerol. As a catalyst to generate unsaturated ketones via Michael addition of β-dicarbonyl compounds.

For general uses, product is also available in powdered form (545856)

Features and Benefits

ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheng Guo et al.
Journal of the American Society for Mass Spectrometry, 23(7), 1191-1201 (2012-05-03)
Collision-induced dissociation (CID) of Li(+) adducts of three sets of compounds that contains an amide bond, including 2-(4, 6-dimethoxypyrimidin-2-ylsulfanyl)-N-phenylbenzamide, its derivatives and simpler structures was investigated by electrospray ionization tandem mass spectrometry (ESI-MS/MS). Observed fragment ions include those that reflect
Manuel Sturzbecher-Höhne et al.
Chemical research in toxicology, 21(12), 2257-2259 (2009-06-24)
We describe the preparation of aqueous solutions of LiONOO and NaONOO from (Me4N)ONOO. An aqueous solution of analytically pure, commercially available (Me4N)ONOO is applied to an Amberlyst 15 column at 4 degrees C, and the Me4N+ is rapidly (in 20
Ana L Aguirre et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 27(51), 12981-12986 (2021-07-08)
High-throughput experimentation (HTE) methods are central to modern medicinal chemistry. While many HTE approaches to C-N and Csp2 -Csp2 bonds are available, options for Csp2 -Csp3 bonds are limited. We report here how the adaptation of nickel-catalyzed cross-electrophile coupling of
Noah P Tu et al.
Angewandte Chemie (International ed. in English), 58(24), 7987-7991 (2019-03-21)
Technologies that enable rapid screening of diverse reaction conditions are of critical importance to methodology development and reaction optimization, especially when molecules of high complexity and scarcity are involved. The lack of a general solid dispensing method for chemical reagents
Anja Kovanda et al.
Scientific reports, 5, 17944-17944 (2015-12-04)
The G4C2 hexanucleotide repeat expansion mutation (HREM) in C9ORF72, represents the most common mutation associated with amyotrophic lateral sclerosis (ALS) and frontotemporal lobar degeneration (FTLD). Three main disease mechanisms have been proposed to date: C9ORF72 haploinsufficiency, RNA toxicity, and accumulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service