Skip to Content
MilliporeSigma
All Photos(4)

Documents

918369

Sigma-Aldrich

4-Aminobenzaldehyde

greener alternative

Synonym(s):

4-Formylaniline, p-Aminobenzaldehyde, p-Formylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7NO
CAS Number:
Molecular Weight:
121.14
Beilstein:
1362885
MDL number:
NACRES:
NA.22

form

solid

Quality Level

greener alternative product score

old score: 30
new score: 21
Find out more about DOZN™ Scoring

greener alternative product characteristics

Atom Economy
Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

2-8°C

InChI

1S/C7H7NO/c8-7-3-1-6(5-9)2-4-7/h1-5H,8H2

InChI key

VATYWCRQDJIRAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of Greener Alternatives. This product belongs to the category of Re-engineered products, showing key improvements in Green Chemistry Principles "Atom Economy", “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstocks”. Click here to view its DOZN scorecard.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service