Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91834

Supelco

BDE No 47

analytical standard

Synonym(s):

2,2′,4,4′-TetraBDE, 2,2′,4,4′-Tetrabromodiphenyl ether, PBDE 47

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H6Br4O
CAS Number:
Molecular Weight:
485.79
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

impurities

≤1.0% water

format

neat

SMILES string

Brc1c(ccc(c1)Br)Oc2c(cc(cc2)Br)Br

InChI

1S/C12H6Br4O/c13-7-1-3-11(9(15)5-7)17-12-4-2-8(14)6-10(12)16/h1-6H

InChI key

XYBSIYMGXVUVGY-UHFFFAOYSA-N

General description

BDE-47 belongs to the class of polybrominated diphenyl ethers (PBDEs), commonly used as fire retardants in consumer products.

Application

BDE-47 may be used as a test material to study its effect on genes involved in thyroid hormone homeostasis of adult female mice via changes in hepatic uridinediphosphate glucuronosyltransferases (UGTs) and transporters.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Possible mechanisms of thyroid hormone disruption in mice by BDE 47, a major polybrominated diphenyl ether congener.
Richardson MV, et al.
Toxicology and Applied Pharmacology, 226(3), 224-250 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service