Skip to Content
MilliporeSigma

916641

2-(3-(Piperidin-4-yl)phenyl)ethan-1-amine dihydrochloride

95%

Synonym(s):

Semi-flexible linker for PROTAC® development

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H22Cl2N2
Molecular Weight:
277.23
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.22
Assay:
95%
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

95%

form

powder

reaction suitability

reagent type: linker

functional group

amine

storage temp.

2-8°C

SMILES string

NCCC1=CC=CC(C2CCNCC2)=C1.Cl.Cl

InChI key

ZFXFMAUVGRWYEM-UHFFFAOYSA-N

Application

2-(3-(Piperidin-4-yl)phenyl)ethan-1-amine dihydrochloride is a 4-aryl piperidine useful as a semi-flexible linker in PROTAC development for targeted protein degradation. Incorporation of rigidity into the linker region of bifunctional protein degraders may impact the 3D orientation of the degrader and thus ternary complex formation as well as optimization of drug-like properties.

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library