Skip to Content
MilliporeSigma
All Photos(1)

Documents

91095

Sigma-Aldrich

Trichloroacetyl isocyanate

purum, ≥97.0% (GC)

Synonym(s):

2,2,2-Trichloroacetyl isocyanate, alpha,alpha,alpha-Trichloroacetyl isocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CCONCO
CAS Number:
Molecular Weight:
188.40
Beilstein:
971201
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.480 (lit.)
n20/D 1.480

bp

80-85 °C/20 mmHg (lit.)

density

1.581 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

ClC(Cl)(Cl)C(=O)N=C=O

InChI

1S/C3Cl3NO2/c4-3(5,6)2(9)7-1-8

InChI key

GRNOZCCBOFGDCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trichloroacetyl isocyanate (TAI) is a versatile building block in organic synthesis. It is also used as an in situ derivatizing reagent for the characterization of phenols, alcohols, and amines.

Application


  • One-pot synthesis of 2-acylaminobenzimidazoles: A study on the synthesis of 2-acylaminobenzimidazoles via a reaction between trichloroacetyl isocyanate and 1,2-phenylenediamine derivatives (Shajari et al., 2018).

  • Cabamothioate compounds: Research on the synthesis of S-aryl (trichloroacetyl) carbamothioate from a reaction of 2-naphthalenethiol or thiophenol derivatives and trichloroacetyl isocyanate (Shajari et al., 2021).

  • Chiral calyx[4]arenes: Diastereoselective synthesis of inherently chiral calyx[4]arenes via reaction of trichloroacetyl isocyanate with 1,3-dihydroxy calixarene (Boyko et al., 2016).

Other Notes

Derivatizing reagent for the structural assignment to hydroxy compounds by NMR; Used in negative ion chemical ionization mass spectrometry

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In situ reactions with trichloroacetyl isocyanate and their application to structural assignment of hydroxy compounds by 1 H NMR spectroscopy. A general comment
Samek et al.
Collection of Czechoslovak Chemical Communications, 44, 558-588 (1979)
D.R. Taylor
Canadian Journal of Chemistry, 54, 189-189 (1976)
H. Fujiwara et al.
Pract. Spectrosc., 3, 329-329 (1980)
Determination of chain branching in epoxy resins by nuclear magnetic resonance spectrometry.
H D Mak et al.
Analytical chemistry, 44(4), 837-839 (1972-04-01)
NMR spectral studies?XII: Trichloroacetyl isocyanate as an in situ derivatizing reagent for 13C NMR spectroscopy of alcohols, phenols and amines
Bose et al.
Tetrahedron, 31, 3025-3029 (1975)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service