Skip to Content
MilliporeSigma

90983

Conjugated (9E,11E)-Linoleic acid

analytical standard

Synonym(s):

(9E,11E)-9,11-Octadecadienoic acid, 9E,11E-CLA, Isolinoleic acid, Linoleic acid (9-trans, 11-trans), Mangold’s acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H32O2
CAS Number:
Molecular Weight:
280.45
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
85151701
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

grade

analytical standard

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

format

neat

functional group

carboxylic acid

storage temp.

−20°C

SMILES string

CCCCCC\C=C\C=C\CCCCCCCC(O)=O

InChI

1S/C18H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h7-10H,2-6,11-17H2,1H3,(H,19,20)/b8-7+,10-9+

InChI key

JBYXPOFIGCOSSB-XBLVEGMJSA-N

General description

Conjugated (9E,11E)-linoleic acid can be obtained via hydrogenation of linoleic acid in the middle [12,13] double bond.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library