Skip to Content
MilliporeSigma

Skip To

900686

Acetonitrile solution

≥99.6%, suitable for mass spectrometry (MS), contains 0.1 % (v/v) formic acid

Synonym(s):

ACN, Acetonitrile with 0.1% formic acid, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
UNSPSC Code:
12191502
NACRES:
NA.03
MDL number:
Beilstein/REAXYS Number:
4124158
Assay:
≥99.6%
Grade:
HPLC grade
Technique(s):
mass spectrometry (MS): suitable
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Acetonitrile solution, contains 0.1 % (v/v) formic acid, for UHPLC, suitable for mass spectrometry (MS)

grade

HPLC grade

Quality Segment

assay

≥99.6%

form

liquid

contains

0.1 % (v/v) formic acid

technique(s)

mass spectrometry (MS): suitable

density

0.782 g/mL at 25 °C

application(s)

food and beverages

format

mixture

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Application


  • Detection of Amino Acids via Chemiluminescence: Acetonitrile is employed in the detection of α-amino acids using peroxyoxalate chemiluminescence. A study demonstrated the use of a mixed solution of water, acetonitrile, and ethyl acetate for detecting amino acids, highlighting its efficacy in analytical applications. This method enhances the sensitivity and accuracy of amino acid detection, making it crucial for biochemical and clinical analyses (Kan et al., American Journal of Analytical Chemistry, 2020).

  • Redox Properties of N-Hydroxyimides: Acetonitrile mixed with sulfuric acid is used to study the redox properties of N-hydroxyimides, acting as a hydrogen atom transfer (HAT) mediator for alcohol oxidation. This application is significant in understanding the electrochemical behavior of organic compounds and developing new catalytic processes for oxidation reactions (Kishioka, Journal of Electroanalytical Chemistry, 2023).

  • Electrode Reactions in Catalytic Mediation: Acetonitrile solution is used in investigating the electrode reactions of N-hydroxyphthalimide in sulfuric acid. This study focuses on its role as a catalytic mediator for alcohol oxidation, providing insights into the electrochemical mechanisms involved. The findings contribute to the development of efficient catalytic systems for organic transformations (Kishioka, Journal of Electroanalytical Chemistry, 2022).

  • Excitation of Europium Complexes: Acetonitrile solution is utilized in the interaction of Eu(FOD)3 with xenon difluoride to study ligand-assisted excitation of Eu(III). This research explores the photophysical properties of europium complexes, which are essential for developing luminescent materials for various applications, including lighting and display technologies (Masyagutova et al., Journal of Fluorine Chemistry, 2024).


Features and Benefits

Acetonitrile solution is used as a solvent for UHPLC-MS with the following advantages:
  • Highest solvent purity for low detection limits.      
  • Extremely low MS baseline noise.        
  • Exemplary lot-to-lot reproducibility.        
  • Microfiltered (0.2 μm).


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
576956574732685461
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

grade

HPLC grade

grade

HPLC grade

grade

HPLC grade

grade

-

technique(s)

mass spectrometry (MS): suitable

technique(s)

HPLC: suitable

technique(s)

HPLC: suitable

technique(s)

HPLC: suitable

assay

≥99.6%

assay

-

assay

-

assay

-

application(s)

food and beverages

application(s)

food and beverages

application(s)

food and beverages

application(s)

food and beverages

form

liquid

form

-

form

liquid

form

liquid


Still not finding the right product?

Explore all of our products under Acetonitrile solution


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

flash_point_f

35.6 °F

flash_point_c

2 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters