Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900637

Sigma-Aldrich

1,2-Dichloroethane

anhydrous, ZerO2®, 99.8%

Synonym(s):

1,2-Dichloroethane ZerO2®, Degassed and low oxygen 1,2-dichloroethane, Ethylene chloride, Ethylene dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2Cl
CAS Number:
Molecular Weight:
98.96
Beilstein:
605264
EC Number:
MDL number:
UNSPSC Code:
12191502
NACRES:
NA.21
grade:
ZerO2®
bp:
83 °C (lit.)
vapor pressure:
87 mmHg ( 25 °C)

grade

ZerO2®

Quality Level

vapor density

3.4 (20 °C, vs air)

vapor pressure

87 mmHg ( 25 °C)

Assay

99.8%

form

liquid

autoignition temp.

775 °F

quality

anhydrous

expl. lim.

15.6 %

dilution

(for analytical testing)

impurities

≤0.003% water
water

refractive index

n20/D 1.444 (lit.)

bp

83 °C (lit.)

mp

−35 °C (lit.)

density

1.256 g/mL at 25 °C (lit.)

SMILES string

ClCCCl

InChI

1S/C2H4Cl2/c3-1-2-4/h1-2H2

InChI key

WSLDOOZREJYCGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

ZerO2 products are rigorously degassed with highly pure inert gas providing solvents and solutions (anhydrous if specified) with very low residual oxygen content.

Legal Information

ZerO2 is a registered trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Asp. Tox. 1 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service