Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
[CH3(CH2)6CH2]4N(NO3)
CAS Number:
Molecular Weight:
528.89
UNSPSC Code:
26111700
NACRES:
NB.61
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
7108220
description
for ion-selective electrodes
Quality Segment
product line
Selectophore™
assay
≥99.0% (NT), ≥99.0%
form
powder
impurities
≤0.05% halides (as bromide)
mp
112-114 °C
SMILES string
[O-][N+]([O-])=O.CCCCCCCC[N+](CCCCCCCC)(CCCCCCCC)CCCCCCCC
InChI
1S/C32H68N.NO3/c1-5-9-13-17-21-25-29-33(30-26-22-18-14-10-6-2,31-27-23-19-15-11-7-3)32-28-24-20-16-12-8-4;2-1(3)4/h5-32H2,1-4H3;/q+1;-1
InChI key
MJHKPBXGJMKYAY-UHFFFAOYSA-N
General description
Tetraoctylammonium nitrate (TOAN) is an ionophore or recognition element mostly used in potentiometric sensors.
Visit our Sensor Applications portal to learn more.
Application
TOAN has been used as recognition element in formulating potentiometric membrane to develop an electronic tongue system for monitoring nitrogen species level in water samples.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Legal Information
Selectophore is a trademark of Merck KGaA, Darmstadt, Germany
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
