Skip to Content
MilliporeSigma
All Photos(1)

Documents

87724

Supelco

Tetramethylammonium bisulfate

suitable for ion pair chromatography, LiChropur, ≥99.0% (T)

Synonym(s):

Tetramethylammonium hydrogen sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)4N(HSO4)
CAS Number:
Molecular Weight:
171.22
Beilstein:
4829202
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NB.21

description

cationic

Quality Level

Assay

≥99.0% (T)

form

crystals

quality

LiChropur

technique(s)

ion pair chromatography: suitable

λ

10 % in H2O

UV absorption

λ: 210 nm Amax: 0.05
λ: 220 nm Amax: 0.04
λ: 230 nm Amax: 0.03
λ: 260 nm Amax: 0.02
λ: 500 nm Amax: 0.02

suitability

corresponds to standard for RP gradient test
corresponds to standard for filter test

SMILES string

C[N+](C)(C)C.OS([O-])(=O)=O

InChI

1S/C4H12N.H2O4S/c2*1-5(2,3)4/h1-4H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

DWTYPCUOWWOADE-UHFFFAOYSA-M

General description

The application of ion-pairing reagents as mobile phase additives ensures the elution of ionic and highly polar substances on reversed phase HPLC columns. The purity of mobile phase additives is of foremost significance for their successful utilization. Sigma-Aldrich offers a huge variety of tailor-made reagents for anionic (quaternary ammonium and phosphonium salts) and cationic (alkanesulfonates) separations. All mobile phase additives are investigated thoroughly with special regard to the requirements of modern reversed phase HPLC:

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sha Liao et al.
Archives of pharmacal research, 37(11), 1416-1425 (2013-12-18)
Chronic hepatitis B virus (HBV) infection may lead to liver cirrhosis and hepatocellular carcinoma, but few drugs are available for its treatment. Acyclic nucleoside phosphonates (ANPs) have remarkable antivirus activities but are not easily absorbed from the gastrointestinal tract and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service