Skip to Content
MilliporeSigma
All Photos(1)

Documents

87350

Sigma-Aldrich

1,2,3,4-Tetrahydroquinoline

purum, ≥96.0% (GC)

Synonym(s):

THQ

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11N
CAS Number:
Molecular Weight:
133.19
Beilstein:
116149
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥96.0% (GC)

form

liquid

refractive index

n20/D 1.593 (lit.)
n20/D 1.594

bp

113-117 °C/10 mmHg (lit.)
249 °C (lit.)

mp

9-14 °C (lit.)

density

1.061 g/mL at 25 °C (lit.)

SMILES string

C1CNc2ccccc2C1

InChI

1S/C9H11N/c1-2-6-9-8(4-1)5-3-7-10-9/h1-2,4,6,10H,3,5,7H2

InChI key

LBUJPTNKIBCYBY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

176.0 °F

Flash Point(C)

80 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arindam Chatterjee et al.
Bioorganic & medicinal chemistry, 22(22), 6409-6421 (2014-12-03)
Calpain mediated cleavage of CDK5 natural precursor p35 causes a stable complex formation of CDK5/p25, which leads to hyperphosphorylation of tau. Thus inhibition of this complex is a viable target for numerous acute and chronic neurodegenerative diseases involving tau protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service