Skip to Content
MilliporeSigma

87296

Tetraheptylammonium bromide

suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
[CH3(CH2)6]4N(Br)
CAS Number:
Molecular Weight:
490.69
UNSPSC Code:
41115711
NACRES:
NB.21
PubChem Substance ID:
EC Number:
224-459-3
Beilstein/REAXYS Number:
3763391
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

cationic

Quality Segment

assay

≥99.0% (AT)

form

flakes

quality

LiChropur

mol wt

490.69 g/mol

technique(s)

ion pair chromatography: suitable

mp

87-89 °C (lit.), 89-91 °C

λ

10 % in methanol

UV absorption

λ: 240 nm Amax: 0.04, λ: 250 nm Amax: 0.03, λ: 260 nm Amax: 0.02, λ: 500 nm Amax: 0.02

suitability

corresponds to standard for filter test

SMILES string

[Br-].CCCCCCC[N+](CCCCCCC)(CCCCCCC)CCCCCCC

InChI

1S/C28H60N.BrH/c1-5-9-13-17-21-25-29(26-22-18-14-10-6-2,27-23-19-15-11-7-3)28-24-20-16-12-8-4;/h5-28H2,1-4H3;1H/q+1;/p-1

InChI key

YQIVQBMEBZGFBY-UHFFFAOYSA-M

Application


  • Novel potentiometric application for the determination of pantoprazole sodium and itopride hydrochloride in their pure and combined dosage form: Highlights a novel potentiometric method where Tetraheptylammonium bromide is used as an ion-pairing agent to enhance the sensitivity and selectivity of drug measurement in pharmaceutical formulations, demonstrating its importance in analytical chemistry and quality control applications (Ragab et al., 2015).

Other Notes

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Tetraheptylammonium bromide


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library