Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

85810

Sigma-Aldrich

STE buffer solution

BioUltra, for molecular biology, pH 7.8

Synonym(s):

TRIS NaCl EDTA buffer solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105319
NACRES:
NA.25

grade

for molecular biology

Quality Level

product line

BioUltra

form

liquid

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

pH

7.8±0.2 (25 °C)

cation traces

Al: ≤1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Li: ≤1 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: 0.03
λ: 280 nm Amax: 0.01

Application


  • Safer DNA extraction from plant tissues using sucrose buffer and glass fiber filter.: The study demonstrates a novel method for DNA extraction from plant tissues using STE buffer solution, ensuring higher safety and efficiency. This approach improves DNA yield and quality, essential for plant genetic research (Takakura et al., 2012).

  • In vitro DNA fragmentation of mitochondrial DNA caused by single-stranded breakage related to macroplasmodial senescence of the true slime mold, Physarum polycephalum.: This study employs STE buffer solution to investigate mitochondrial DNA fragmentation in the slime mold Physarum polycephalum, providing valuable insights into cellular aging processes (Abe et al., 2000).

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service