Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

85431

Supelco

Silylating mixture I according to Sweeley

for GC derivatization, LiChropur

Synonym(s):

Chlorotrimethylsilane – hexamethyldisilazane mixture, Sylon HTP, hexamethyldisilazane/chlorotrimethylsilane/pyridine = 2:1:10 (v/v/v), HMDS + TMCS

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for GC derivatization

Quality Level

description

ready to use solution in pyridine (10 vol parts)

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

technique(s)

gas chromatography (GC): suitable

storage temp.

2-8°C

SMILES string

C[Si](C)(C)Cl.C[Si](C)(C)N[Si](C)(C)C.C[Si](C)(C)N[Si](C)(C)C

InChI

1S/2C6H19NSi2.C3H9ClSi/c2*1-8(2,3)7-9(4,5)6;1-5(2,3)4/h2*7H,1-6H3;1-3H3

InChI key

UCJNZOHHMSKSRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Learn more in the Product Information

Components

hexamethyldisilazane : chlorotrimethylsilane : pyridine = 2:1:10 (v/v)

Other Notes

Useful silylating mixture for the silylation of hydroxy-compounds

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany
Sylon is a trademark of Sigma-Aldrich Co. LLC

related product

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

J. Heberle et al.
Silylating Agents, 2nd ed. (1995)
Sweeley, C.C., et al.
Journal of the American Chemical Society, 85, 2497-2497 (1963)

Articles

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service