Skip to Content
MilliporeSigma

85265

Methyl 4-hydroxybenzoate sodium salt

tested according to Ph. Eur.

Synonym(s):

Methylis parahydroxybenzoas natricum, Sodium methyl 4-hydroxybenzoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C8H7NaO3
CAS Number:
Molecular Weight:
174.13
UNSPSC Code:
12352300
NACRES:
NA.21
PubChem Substance ID:
EC Number:
225-714-1
Beilstein/REAXYS Number:
4165000
MDL number:
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

tested according to Ph. Eur.

form

powder

application(s)

research use (small molecule)

SMILES string

[Na+].COC(=O)c1ccc([O-])cc1

InChI

1S/C8H8O3.Na/c1-11-8(10)6-2-4-7(9)5-3-6;/h2-5,9H,1H3;/q;+1/p-1

InChI key

PESXGULMKCKJCC-UHFFFAOYSA-M

Application


  • Gellan gum gel tissue phantoms and gel dosimeters with tunable electrical, mechanical and dosimetric properties.: This study utilizes methyl 4-hydroxybenzoate sodium salt in the preparation of gellan gum gel tissue phantoms. The research highlights its role in enhancing the electrical, mechanical, and dosimetric properties of the gels, making them suitable for various biomedical applications such as tissue phantoms and dosimeters (Brzozowski P et al., 2021).

  • Ultrasound-Assisted Surfactant-Enhanced Emulsification Micro-Extraction Followed by HPLC for Determination of Preservatives in Water, Beverages and Personal Care Products.: The research highlights the application of methyl 4-hydroxybenzoate sodium salt in ultrasound-assisted surfactant-enhanced emulsification micro-extraction, demonstrating its utility in the extraction and analysis of preservatives from various matrices (Jan-E S et al., 2017).



Still not finding the right product?


pictograms

Corrosion

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library