Skip to Content
MilliporeSigma
All Photos(1)

Documents

83650

Millipore

L-Rhamnose monohydrate

suitable for microbiology, ≥99.0%,

Naturally occurring deoxy sugar that is found primarily in plants and some bacteria

Synonym(s):

6-Deoxy-L-mannose monohydrate, L(+)Rhamnopyranose, 6-Deoxy-L-mannose, L(+)Rhamnopyranose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O5 · H2O
CAS Number:
Molecular Weight:
182.17
Beilstein:
3912643
EC Number:
MDL number:
UNSPSC Code:
41106212
PubChem Substance ID:
NACRES:
NA.85

Quality Level

Assay

≥99.0% (HPLC)
≥99.0%

form

powder

optical activity

[α]20/D +8±0.5°, 2 hr, c = 5% in H2O

ign. residue

≤0.1% (as SO4)

mp

90-95 °C

solubility

H2O: 0.1 g/mL, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

As: ≤0.1 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

application(s)

microbiology

storage temp.

10-30°C

SMILES string

O.C[C@@H]1O[C@@H](O)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C6H12O5.H2O/c1-2-3(7)4(8)5(9)6(10)11-2;/h2-10H,1H3;1H2/t2-,3-,4+,5+,6+;/m0./s1

InChI key

BNRKZHXOBMEUGK-NRBMBCGPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Rhamnose is a deoxy sugar and got its name from the plant Buckthorn (Latin: Rhamnus) where it was isolated. The L-Rhamnose is the naturally predominant sugar form. In nature the rhamnose can be found, additionally to Buckthorn in poison Sumac, plants of the genus Uncaria and in microalgae belonging to class Bacillariophyceae (diatoms).In nature rhamnose is often found linked to other sugars. It is a common component of glycosides from many plants and found in the peptidoglycan of the cell wall of acid-fast bacteria in the Mycobacterium genus.
Rhamnose can also be fermented by several microorganisms like E. coli, Enterobacter, Citrobacter, Klebsiella and some species of Salmonella. Acid production from rhamnose is a characteristic phenotype of Listeria monocytogenes. Therefore, rhamnose can be used for differentiation and identification of bacteria, it is used in media like Rhamnose broth or Phenol red Rhamnose broth.

Application

L-Rhamnose monohydrate is used in many media formulations such as Rhamnose broth or Phenol red Rhamnose broth to facilitate differentiation of bacteria that can ferment the sugar and those that cannot.
An understanding of the rhamnose-containing polysaccharides present in the cell walls of certain bacteria can enable us to identify biosynthetic pathways that can further be used as potential targets for antibacterial therapy.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pooja Flora et al.
Cell reports, 25(13), 3828-3843 (2018-12-28)
Maternal mRNAs synthesized during oogenesis initiate the development of future generations. Some maternal mRNAs are either somatic or germline determinants and must be translationally repressed until embryogenesis. However, the translational repressors themselves are temporally regulated. We used polar granule component
Ivana R Sequeira et al.
Clinical and experimental pharmacology & physiology, 41(2), 107-117 (2013-09-17)
The effects of inflammatory changes on the absorption of different-sized probes and their permeability ratios are poorly understood. The aim of the present study was to determine the effects of a pharmacological agent on the permeability of the gut mucosa

Articles

Culture media provides a habitat with suitable nutrients, energy sources, and certain environmental conditions for the growth of microorganisms. The components of the culture media range from simple sugars to peptones, salts, antibiotics, and complex indicators.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service