Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

81172

Sigma-Aldrich

Poly(ethylene glycol)

tested according to Ph. Eur., 400

Synonym(s):

Macrogol 400, Polyethylene glycol, PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

Agency

tested according to Ph. Eur.

Quality Level

form

liquid

mol wt

380-420

application(s)

pharmaceutical (small molecule)

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polyethylene glycol (PEG) is a polyether compound[1] with repeated ethylene glycol units[-(CH2CH2O)n].[2]

Application

Poly(ethylene glycol) has been used as a liquid medium to prepare shear thickening fluid (STF) by directly mixing it with fumica silica.[3][4][5]

Biochem/physiol Actions

Polyethylene glycol (PEG) is a synthetic polymer[6] and is used as an additive in food, cosmetics, and pharmaceuticals.[1] It is also involved in biomedical applications like biosensing, imaging, and tissue engineering. PEG is used as a gold standard in bioconjugation and nanomedicine thereby increasing blood circulation time and improving drug efficacy. PEG-conjugated drugs are approved by the U.S. Food and Drug Administration (FDA). They are considered safe for use in humans.[6] PEG reduces the liver uptake and repels protein adsorption, which makes it useful in nanocoating.[7]

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The rheology of shear thickening fluids with various ceramic particle additives
Gurgen S, et al.
Materials & Design, 104, 312-319 (2016)
The ballistic performance of aramid based fabrics impregnated with multi-phase shear thickening fluids
Gurgen S and Kucshan MC
Polymer Testing, 64, 296-306 (2017)
The stab resistance of fabrics impregnated with shear thickening fluids including various particle size of additives
Gurgen S and Kucshan MC
Composites Part A: Applied Science and Manufacturing, 94, 50-60 (2017)
Anisha A D'souza et al.
Expert opinion on drug delivery, 13(9), 1257-1275 (2016-04-28)
Polyethylene glycol (PEG) is a polymer of choice in drug delivery systems. This USFDA-approved polymer is popular due to its tunable properties and well-established safety profile: prime requisites considered during the selection of any excipient in formulation development. The unique
Janina-Miriam Noy et al.
Biomacromolecules, 21(6), 2320-2333 (2020-04-29)
Phosphorylcholine is known to repel the absorption of proteins onto surfaces, which can prevent the formation of a protein corona on the surface of nanoparticles. This can influence the fate of nanoparticles used for drug delivery. This material could therefore

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service