Skip to Content
MilliporeSigma
All Photos(1)

Documents

80816

Supelco

(2S)-2-(5-Chloro-6-methoxynaphthalen-2-yl)propanoic acid

pharmaceutical impurity standard

Synonym(s):

Naproxen Impurity B, (αS)-5-Chloro-6-methoxy-α-methyl-2-naphthaleneacetic acid, (2S)-2-(5-Chloro-6-methoxy-2-naphthyl)propionic acid, (S)-5-Chloronaproxen

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H13ClO3
CAS Number:
Molecular Weight:
264.70
Beilstein:
5278973
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

Quality Level

API family

naproxen

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤0.5% water

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C[C@H](C(O)=O)C1=CC2=CC=C(OC)C(Cl)=C2C=C1

InChI

1S/C14H13ClO3/c1-8(14(16)17)9-3-5-11-10(7-9)4-6-12(18-2)13(11)15/h3-8H,1-2H3,(H,16,17)/t8-/m0/s1

InChI key

RVVAYDHIVLCPBC-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

related product

Product No.
Description
Pricing

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service