Skip to Content
MilliporeSigma
All Photos(1)

Documents

80704

Millipore

VCC Selective Supplement

suitable for microbiology

Synonym(s):

ECO157:H7 Selective Supplement, Vancomycin Cefixime Cefsulodin Selective Supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171614
NACRES:
NA.85

sterility

sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental
food and beverages
pathogen testing

microbiology

storage temp.

2-8°C

suitability

Escherichia coli
coliforms

Application

VCC Selective Supplement is an antibiotic supplement recommended for selective isolation of Escherichia coli O157:H7. It has been used to study the effects of adding VCC on enrichment time using heart infusion broth, tryptic soy broth (TSB) and buffered peptone water.

Components

(per vial, sufficient for 1000 ml medium)
Vancomycin 8.0 mg
Cefixime 0.05 mg
Cefsoludin 10.0 mg

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiplex real-time PCR assays for detection of eight Shiga toxin-producing Escherichia coli in food samples by melting curve analysis
Singh, Prashant
International Journal of Food Microbiology, 101-108 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service