Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

80605

Supelco

(+)-α-Pinene

analytical standard

Synonym(s):

(1R,5R)-2,6,6-Trimethylbicyclo[3.1.1]hept-2-ene, (1R,5R)-2-Pinene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
Beilstein:
2038653
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (sum of enantiomers, GC)

optical activity

[α]20/D +51±2°, neat

optical purity

enantiomeric ratio: ≥98:2 (GC)

autoignition temp.

491 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.465 (lit.)

bp

155-156 °C (lit.)

mp

−62 °C (lit.)

density

0.858 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CC1=CC[C@@H]2C[C@H]1C2(C)C

InChI

1S/C10H16/c1-7-4-5-8-6-9(7)10(8,2)3/h4,8-9H,5-6H2,1-3H3/t8-,9-/m1/s1

InChI key

GRWFGVWFFZKLTI-RKDXNWHRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(+)-α-Pinene is a monoterpenoid compound, which comprises of an endocyclic double bond and is found in the resins of pine trees and other conifers.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Justin J Miller et al.
ACS infectious diseases, 6(10), 2567-2572 (2020-09-24)
Infection with malarial parasites renders hosts more mosquito-attractive than their uninfected, healthy counterparts. One volatile organic compound, α-pinene, is associated with Plasmodium spp. infection in multiple studies and is a known mosquito attractant. However, how malarial infection results in elevated
Seinfeld.HJ and Pandis.NS et al.
Atmospheric Chemistry and Physics: From Air Pollution to Climate Change, (3) (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service