Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
HOC6H4CO2C2H5
CAS Number:
Molecular Weight:
166.17
EC Number:
204-399-4
UNSPSC Code:
41116107
NACRES:
NA.24
Beilstein/REAXYS Number:
1101972
MDL number:
eCl@ss:
39024522
Quality Segment
product line
TraceCERT®
grade
certified reference material, TraceCERT®
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
technique(s)
gas chromatography (GC): suitable
bp
297-298 °C (lit.)
mp
114-117 °C (lit.)
application(s)
cosmetics
food and beverages
format
neat
SMILES string
CCOC(=O)c1ccc(O)cc1
InChI
1S/C9H10O3/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6,10H,2H2,1H3
InChI key
NUVBSKCKDOMJSU-UHFFFAOYSA-N
General description
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.
Find all available reference materials for compounds listed in 10/2011 here
Find all available reference materials for compounds listed in 10/2011 here
Legal Information
TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Explore all of our products under Ethylparaben
Storage Class
11 - Combustible Solids
WGK
WGK 1
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.