Skip to Content
MilliporeSigma

Skip To

795461

Ammonium phosphate monobasic

anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥98%

Synonym(s):

mono-Ammonium phosphate, prim-Ammonium phosphate, Ammonium dihydrogenphosphate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
NH4H2PO4
CAS Number:
Molecular Weight:
115.03
NACRES:
NA.21
PubChem Substance ID:
UNSPSC Code:
12352300
EC Number:
231-764-5
MDL number:
Assay:
≥98%
Grade:
ACS reagent, anhydrous
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

ACS reagent, anhydrous

vapor pressure

<0 hPa ( 20 °C)

product line

Redi-Dri

assay

≥98%

form

powder

quality

free-flowing

impurities

≤0.005% NH4OH pptd., ≤0.005% insolubles

pH

3.8-4.4 (25 °C, 5%)

mp

190 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤5 ppm, nitrate (NO3-): ≤0.001%, sulfate (SO42-): ≤0.01%

cation traces

Ca: 0.001%, Fe: ≤0.001%, K: ≤0.005%, Mg: 0.005%, Na: ≤0.005%, heavy metals (as Pb): ≤5 ppm

SMILES string

N.OP(O)(O)=O

InChI

1S/H3N.H3O4P/c;1-5(2,3)4/h1H3;(H3,1,2,3,4)

InChI key

LFVGISIMTYGQHF-UHFFFAOYSA-N

General description

Ammonium dihydrogen phosphate (ADP) is a crystal material and is a member of large family of compounds which possess a chemical formula:MH2XO4 .

Application


  • Capillary-seeding crystallization and preliminary crystallographic analysis of a solvent-tolerant elastase from Pseudomonas aeruginosa strain K.: This research employs ammonium phosphate monobasic in the crystallization process of elastase from Pseudomonas aeruginosa. The study provides crucial structural data that can aid in understanding the enzyme′s function and its potential industrial applications (Ali et al., 2013).



Biochem/physiol Actions

Ammonium dihydrogenphosphate (ADP) exhibits non-linear optical (NLO) and electro-optic (EO) properties.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A16450970901-4970
description

anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥98%

description

ReagentPlus®, ≥98.5%

description

BioUltra, ≥99.5% (T)

description

SAJ first grade, ≥98.0%

form

powder

form

powder

form

-

form

crystalline

assay

≥98%

assay

≥98.5%

assay

≥99.5% (T)

assay

≥98.0%

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

-

grade

ACS reagent, anhydrous

grade

-

grade

-

grade

SAJ first grade

impurities

≤0.005% NH4OH pptd., ≤0.005% insolubles

impurities

-

impurities

insoluble matter, passes filter test

impurities

-

mp

190 °C (dec.) (lit.)

mp

190 °C (dec.) (lit.)

mp

190 °C (dec.) (lit.)

mp

190 °C (dec.) (lit.)


Still not finding the right product?

Explore all of our products under Ammonium phosphate monobasic


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters